Order Entry
Denmark
Orders LinkContactUsLinkComponent
Tripod stands
Tripod stands
Supplier:   USBECK Laborgeräte
Tripod stands
Certificates
Tripod stands
2 Options Available Below
Product Details & Documents
Retort stand base in tripod shape made from powder-coated steel.
  • For rods up to 13 mm Ø
  • With hinged screw M 6
Thickness: 10 mm
Product Family Options
Product Information
  • Weight
    Arm length
    Availability
    Price
  • USBE2031
    Retort stand base, tripod
    Retort stand base, tripod
    1,1 kg
    115 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 381,00 kr
    381,00 kr
    1 * 1 Items
       381,00 kr
  • USBE2033
    Retort stand base, tripod
    Retort stand base, tripod
    1,7 kg
    185 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 522,00 kr
    522,00 kr
    1 * 1 Items
       522,00 kr