Safety spectacles, Prism
Anti-UV, anti-scratch, anti-fog, Transparent, Clear PC
Catalog # BOLEPRIPSI
Supplier: Bollé Safety
Jump to:
Product Details & Documents
The model Prism with dual lens offers a high level of protection and its frame is entirely composed of ultra resistant PC.
- Panoramic vision
- Sporty design and provides upper protection
- Comfortable with straight temples
Weight: 23 g
Certifications: EN 166; EN 170
Delivery information: Supplied with adjustable cord.
Specifications
- Type:Anti-UV, anti-scratch, anti-fog
- Colour:Transparent
- Lenses:Clear PC