Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
Synonyms: Methyl 2-cyclopentanonecarboxylate
:
:142.16 g/mol
EINECS:233-962-7
CAS Number:10472-24-9
Formula:C₇H₁₀O₃
MDL Number:MFCD00001411