Ethyl-2-oxocyclopentanecarboxylate 95+%
:611-10-9
:
:C₈H₁₂O₃
:MFCD00001412
:224…228 °C (760 mmHg)
:1.05 g/cm³
:77 °C
:156.18 g/mol
:Refrigerator
:210-253-0
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4500 to 1.4540 (20°C, 589 nm)
- GC:≥95.0 %
Frequently Bought Together