
Methyl-3-(bromomethyl)benzoate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1129-28-8
:
Formula:C₉H₉BrO₂
MDL Number:MFCD00051437
Boiling Pt:115 °C (0.8 mmHg)
Melting Pt:43…45 °C
EINECS:419-100-4
ADR:8,III
:229.07 g/mol
:GHS05
Storage Temperature:Ambient
Related Information
Frequently Bought Together