
1-Naphthyl phosphate monosodium salt monohydrate ≥97.0% (by titrimetric analysis) substrate for Phosphatase
Supplier: TCI
Certificates
About this item
CAS Number:81012-89-7
:
Formula:C₁₀H₈NaO₄P·1H₂O
MDL Number:MFCD00041008
:264.15 g/mol
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (Nonaqueous Titration):min. 97.0 %
- Water:5.3 to 8.3 %
- Alkaline Phosphatase detection test:to pass test
Specification Test Results
Frequently Bought Together