
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12) ≥98.0% (by HPLC, total nitrogen) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:14933-08-5
:
Formula:C₁₇H₃₇NO₃S
MDL Number:MFCD00036909
:335.55 g/mol
Melting Pt:244°C
Storage Temperature:Ambient
EINECS:239-002-3
ADR:8,II
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC) (CAD):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Suitability for protein-analysis:to pass test
Specification Test Results
Frequently Bought Together