Order Entry
Denmark
ContactUsLinkComponent
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12) ≥98.0% (by total nitrogen basis)
Irritant
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12) ≥98.0% (by total nitrogen basis)
Supplier:  TCI
Certificates
undefined
Irritant
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12) ≥98.0% (by total nitrogen basis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Solubility in Water:
    almost transparency

Specification Test Results

Frequently Bought Together