


Bis(cyclopentadienyl)zirconium hydridochloride ≥96.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:37342-97-5
:
Formula:C₁₀H₁₁ClZr
MDL Number:MFCD02089401
:257.87 g/mol
Merck Index:14,08396
EINECS:253-479-5
UN:3131
ADR:4.3,II
:GHS02
:GHS05
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (Chelometric Titration):min. 96.0 %
Specification Test Results
Frequently Bought Together