Order Entry
Denmark
ContactUsLinkComponent
2,2,4-Trimethylpentane ≥99.0% (by GC) for HPLC
Flammable
Irritant
Health
Environment
2,2,4-Trimethylpentane ≥99.0% (by GC) for HPLC
Catalog # TCIAI0276-500ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
Health
Environment
2,2,4-Trimethylpentane ≥99.0% (by GC) for HPLC
Catalog # TCIAI0276-500ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.0 %
  • Absorbance (210nm):
    max. 0.500
  • Absorbance (220nm):
    max. 0.200
  • Absorbance (230nm):
    max. 0.080

Specification Test Results

About this item

 :
 :114.23 g/mol
Merck Index:14,05193
Boiling Pt:99°C
Melting Pt:–107 °C
Density:0,692 g/cm³ (20 °C)
Flash Pt:-9°C
Flash Pt:–9°C
Storage Temperature:Ambient
EINECS:208-759-1
UN:1262
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
 :GHS09
CAS Number:540-84-1
Formula:(CH₃)₂CHCH₂C(CH₃)₃
MDL Number:MFCD00008943