Order Entry
Denmark
ContactUsLinkComponent
 
 
 
 
Lead tetraacetate ≥96.0% (by titrimetric analysis) (contains Acetic Acid)
 :  TCI
 
undefined
 
 
 
Lead tetraacetate ≥96.0% (by titrimetric analysis) (contains Acetic Acid)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Iodometric Titration):
    min. 96.0 %

 

Frequently Bought Together