Tetrazolium blue chloride ≥95.0% (by titrimetric analysis) for biochemical research
: TCI
:1871-22-3
:
:C₄₀H₃₂Cl₂N₈O₂
:MFCD00040933
:14,09244
:248°C
:727.65 g/mol
:Ambient
:217-488-8
- Appearance:White to Amber powder to crystal
- Purity (HPLC):min. 90.0 area%
- Purity (Nonaqueous Titration):min. 95.0 % (calcd.on dried substance)
- Drying loss:max. 3.0 %
- Alkaline Phosphatase detection test:to pass test
Frequently Bought Together