Order Entry
Denmark
ContactUsLinkComponent
 
 
 
 
 
2-Methylstyrene ≥97.0% (by GC) stabilised
  TCIAM0259-5ML
 :  TCI
 :  
undefined
 
 
 
 
2-Methylstyrene ≥97.0% (by GC) stabilised
  TCIAM0259-5ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 97.0 %

 

 

 :
 :118.18 g/mol
 :171°C
 :-69°C
 :–69°C
 :0,9284 g/cm³ (0 °C)
 :60°C
 :Refrigerator
 :210-256-7
 :2618
 :3,III
 :Stabilised with 4-tert-butylpyrocatechol (TBC)
 :GHS02
 :GHS07
 :GHS08
 :GHS09
 :611-15-4
 :C₉H₁₀
 :MFCD00014936