Order Entry
Denmark
ContactUsLinkComponent
Dodecyltrimethylammonium chloride ≥98.0% (by titrimetric analysis) for ion pair chromatography
Irritant
Environment
Dodecyltrimethylammonium chloride ≥98.0% (by titrimetric analysis) for ion pair chromatography
Supplier:  TCI
Certificates
undefined
Irritant
Environment
Dodecyltrimethylammonium chloride ≥98.0% (by titrimetric analysis) for ion pair chromatography
Supplier:  TCI

Specification Test Results

  • Appearance:
    White powder to crystal
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Absorbance (210nm):
    max. 0.200
  • Absorbance (230nm):
    max. 0.070
  • Absorbance (254nm):
    max. 0.040

Specification Test Results

Related Information

Frequently Bought Together