D-(+)-Trehalose, anhydrous ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:99-20-7
:
Formula:C₁₂H₂₂O₁₁
MDL Number:MFCD00006628
:342.3 g/mol
Merck Index:14,09580
Melting Pt:203 °C
Density:1,53 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:202-739-6
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Specific rotation [a]20/D:+194.0 to +200.0 deg (C=7, H2O)
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together