Methyl-4-hydroxybenzoate ≥99.0% (by HPLC, titration analysis) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:99-76-3
:
Formula:HOC₆H₄CO₂CH₃
MDL Number:MFCD00002352
:152.15 g/mol
Merck Index:14,06107
Boiling Pt:280°C
Melting Pt:125…128 °C
Density:1,1208 g/cm³ (137 °C)
EINECS:202-785-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Neutralization titration):min. 99.0 %
- Melting point:125.0 to 128.0 deg-C
- Protein impurity test (SDS-PAGE):to pass test
Specification Test Results
Frequently Bought Together