
1H,1H,2H,2H-Perfluoro-1-iodohexane ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2043-55-2
:
Formula:C₆H₄F₉I
MDL Number:MFCD00039409
:373.99 g/mol
Boiling Pt:126°C
EINECS:218-055-6
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow to Light red clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together