Ordreindtastning
Denmark
ContactUsLinkComponent
N,N,N',N'-Tetramethylethylendiamin ≥98.0% (by GC, titration analysis) til elektroforese
N,N,N',N'-Tetramethylethylendiamin ≥98.0% (by GC, titration analysis) til elektroforese
Supplier:  TCI
Certificates
undefined
N,N,N',N'-Tetramethylethylendiamin ≥98.0% (by GC, titration analysis) til elektroforese
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Refractive index n20/D:
    1.4150 to 1.4190
  • Functionality test:
    to pass test

Specification Test Results

Related Information

Frequently Bought Together