


Ammonium peroxodisulphate (APS) ≥99.0% (by titrimetric analysis) for electrophoresis
Supplier: TCI
Certificates
About this item
CAS Number:7727-54-0
:
Formula:(NH₄)₂S₂O₈
MDL Number:MFCD00003390
:228.2 g/mol
Merck Index:14,00541
Melting Pt:120°C
Density:1,98 g/cm³ (20 °C)
EINECS:231-786-5
UN:1444
ADR:5.1,III
:GHS03
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White powder to crystal
- Purity (Potassium permanganate method):min. 99.0 %
- Functionality test:to pass test
Specification Test Results
Frequently Bought Together