Order Entry
Denmark
ContactUsLinkComponent
Isoeugenol cis- and trans-mixture ≥97.0% (by GC)
Irritant
Health
Isoeugenol cis- and trans-mixture ≥97.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
Health
Isoeugenol cis- and trans-mixture ≥97.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Yellow to Green clear liquid
  • Purity (GC):
    min. 98.0 % (total of isomer)

Specification Test Results

Related Information

Frequently Bought Together