Test tubes, with rim, medium wall
Supplier: MURRAY S
Certificates
About this item
Neutral glass, SAMCO '50'
- Medium walled
- Withstand moderate temperature change
Certifications: Manufactured to the dimensions, tolerances and quality requirements of ISO 4142:2002, to meet all applications for re-usable glass tubes.
Caution: Tubes should not be taken from cold and placed directly into the hottest part of the flame without pre-heating.
Frequently Bought Together