Order Entry
Denmark
ContactUsLinkComponent
Cation exchange resins, Toyopearl® and TSK-GEL®
Cation exchange resins, Toyopearl® and TSK-GEL®
Supplier:  TOSOH Bioscience
Certificates
undefined
Cation exchange resins, Toyopearl® and TSK-GEL®
Supplier:  TOSOH Bioscience

Specifications

  • Amount of ionic groups:
    60 - 120 µeq/ml
  • Average particle size:
    15 - 25 µm
  • Pressure resistance:
    3 bar
  • Working pH:
    2-13

Specifications

Frequently Bought Together