Order Entry
Denmark
ContactUsLinkComponent
Pipette tips for Eppendorf®
Pipette tips for Eppendorf®
  GREI765280UK
undefined
Pipette tips for Eppendorf®
  GREI765280UK
  • Packed:
    96 per ST Rack (yellow)
  • Sterile:
    -
  • Volume:
    0,5 - 20 µl
  • Length:
    45,6 mm
  • Seq:
    04
  • Environmentally Preferable:
  • Tip style:
    Micro, crystal tip, UK
  • Tip colour:
    Transparent
  • For use with:
    Eppendorf® 4710, Reference, Research Pro
  • Pk:
    960
These pipette tips feature a shorter tip collar and are recommended for use with Eppendorf® pipettors.
  • Bevelled edge increases pipetting accuracy
  • Manufactured from high-grade PP to give the optimum in performance and fit
  • Free of heavy metals
  • Autoclavable
 : Matching racks for bulkware tips can be ordered separately.