Kieselguhr filter papers
Supplier: MACHEREY-NAGEL
About this item
This filter paper retains very fine turbidities and is e.g. recommended for the clarification of urines or sugar solutions.
13 Options Available Below
Product Details & Documents
This filter paper retains very fine turbidities and is e.g. recommended for the clarification of urines or sugar solutions.
Specifications for Product Family
Specifications
- Catalog No:
- MANA447009
- MANA447011
- MANA447012
- MANA447015
- MANA447018
- MANA447024
- MANA447032
- MANA547011
- MANA547012
- MANA547015
- MANA547018
- MANA547024
- MANA547032
- Weight:
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- 140 g/m²
- Thickness:
- 0,32 mm
- 0,32 mm
- 0,32 mm
- 0,32 mm
- 0,32 mm
- 0,32 mm
- 0,32 mm
- 0,32 mm
- 0,32 mm
- 0,32 mm
- 0,32 mm
- 0,32 mm
- 0,32 mm
- Filtration speed:
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)
Specifications
Product Family Options
Grade MN 660
Product Information
- GradeSizeAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.MANA447009Weight140 g/m²Thickness0,32 mmFiltration speed50 s (Herzberg) - Specifications:
More
Specifications
Cat. No.MANA447011Weight140 g/m²Thickness0,32 mmFiltration speed50 s (Herzberg) - Specifications:
More
Specifications
Cat. No.MANA447012Weight140 g/m²Thickness0,32 mmFiltration speed50 s (Herzberg) - Specifications:
More
Specifications
Cat. No.MANA447015Weight140 g/m²Thickness0,32 mmFiltration speed50 s (Herzberg) - Specifications:
More
Specifications
Cat. No.MANA447018Weight140 g/m²Thickness0,32 mmFiltration speed50 s (Herzberg) - Specifications:
More
Specifications
Cat. No.MANA447024Weight140 g/m²Thickness0,32 mmFiltration speed50 s (Herzberg) - Specifications:
More
Specifications
Cat. No.MANA447032Weight140 g/m²Thickness0,32 mmFiltration speed50 s (Herzberg)
Grade MN 660, folded
Product Information
- GradeSizeAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.MANA547011Weight140 g/m²Thickness0,32 mmFiltration speed50 s (Herzberg) - Specifications:
More
Specifications
Cat. No.MANA547012Weight140 g/m²Thickness0,32 mmFiltration speed50 s (Herzberg) - Specifications:
More
Specifications
Cat. No.MANA547015Weight140 g/m²Thickness0,32 mmFiltration speed50 s (Herzberg) - Specifications:
More
Specifications
Cat. No.MANA547018Weight140 g/m²Thickness0,32 mmFiltration speed50 s (Herzberg) - Specifications:
More
Specifications
Cat. No.MANA547024Weight140 g/m²Thickness0,32 mmFiltration speed50 s (Herzberg) - Specifications:
More
Specifications
Cat. No.MANA547032Weight140 g/m²Thickness0,32 mmFiltration speed50 s (Herzberg)