Order Entry
Denmark
Orders LinkContactUsLinkComponent
Filter papers for fat analysis
Filter papers for fat analysis
Filter papers for fat analysis
Filter papers for fat analysis
Smooth folded filter papers for fat analysis.
MN 615 ff paper is particularly suited for the analysis of fats. A special treatment with organic solvents guarantees that these filters are practically free of fats and resins (ether-soluble residue < 0,1 mg for a 270 mm filter circle).
MN 715 paper is also suited for the analysis of fats. Careful selection of the raw materials ensures a low ether-soluble residue for these filters.
  • Catalog No:
  • MANA591005
  • MANA591007
  • MANA591009
  • MANA591011
  • MANA591012
  • MANA591015
  • MANA591018
  • MANA591024
  • MANA591027
  • MANA528005
  • MANA528007
  • MANA528009
  • MANA528011
  • MANA528012
  • MANA528015
  • MANA528018
  • MANA528024
  • MANA528027
  • MANA561032
  • Weight:
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • Thickness:
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • Filtration speed:
  • 22 s
  • 22 s
  • 22 s
  • 22 s
  • 22 s
  • 22 s
  • 22 s
  • 22 s
  • 22 s
  • 22 s
  • 22 s
  • 22 s
  • 22 s
  • 22 s
  • 22 s
  • 22 s
  • 22 s
  • 22 s
  • 22 s
Product Family Options
  • Grade
    Size
  • MN 615 ff
    55 mm
    1 * 100 Items
    1 * 100 Items - 742,00 kr
    742,00 kr
    1 * 100 Items
       742,00 kr
    Specifications
    Cat. No.
    MANA591005
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Filtration speed
    22 s
  • MN 615 ff
    70 mm
    1 * 100 Items
    1 * 100 Items - 742,00 kr
    742,00 kr
    1 * 100 Items
       742,00 kr
    Specifications
    Cat. No.
    MANA591007
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Filtration speed
    22 s
  • MN 615 ff
    90 mm
    1 * 100 Items
    1 * 100 Items - 742,00 kr
    742,00 kr
    1 * 100 Items
       742,00 kr
    Specifications
    Cat. No.
    MANA591009
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Filtration speed
    22 s
  • MN 615 ff
    110 mm
    1 * 100 Items
    1 * 100 Items - 742,00 kr
    742,00 kr
    1 * 100 Items
       742,00 kr
    Specifications
    Cat. No.
    MANA591011
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Filtration speed
    22 s
  • MN 615 ff
    125 mm
    1 * 100 Items
    1 * 100 Items - 742,00 kr
    742,00 kr
    1 * 100 Items
       742,00 kr
    Specifications
    Cat. No.
    MANA591012
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Filtration speed
    22 s
  • MN 615 ff
    150 mm
    1 * 100 Items
    1 * 100 Items - 742,00 kr
    742,00 kr
    1 * 100 Items
       742,00 kr
    Specifications
    Cat. No.
    MANA591015
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Filtration speed
    22 s
  • MN 615 ff
    185 mm
    1 * 100 Items
    1 * 100 Items - 839,00 kr
    839,00 kr
    1 * 100 Items
       839,00 kr
    Specifications
    Cat. No.
    MANA591018
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Filtration speed
    22 s
  • MN 615 ff
    240 mm
    1 * 100 Items
    1 * 100 Items - 1.350,00 kr
    1.350,00 kr
    1 * 100 Items
       1.350,00 kr
    Specifications
    Cat. No.
    MANA591024
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Filtration speed
    22 s
  • MN 615 ff
    270 mm
    1 * 100 Items
    1 * 100 Items - 1.550,00 kr
    1.550,00 kr
    1 * 100 Items
       1.550,00 kr
    Specifications
    Cat. No.
    MANA591027
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Filtration speed
    22 s
  • MN 715
    55 mm
    1 * 100 Items
    1 * 100 Items - 311,00 kr
    311,00 kr
    1 * 100 Items
       311,00 kr
    Specifications
    Cat. No.
    MANA528005
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Filtration speed
    22 s
  • MN 715
    70 mm
    1 * 100 Items
    1 * 100 Items - 311,00 kr
    311,00 kr
    1 * 100 Items
       311,00 kr
    Specifications
    Cat. No.
    MANA528007
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Filtration speed
    22 s
  • MN 715
    90 mm
    1 * 100 Items
    1 * 100 Items - 311,00 kr
    311,00 kr
    1 * 100 Items
       311,00 kr
    Specifications
    Cat. No.
    MANA528009
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Filtration speed
    22 s
  • MN 715
    110 mm
    1 * 100 Items
    1 * 100 Items - 311,00 kr
    311,00 kr
    1 * 100 Items
       311,00 kr
    Specifications
    Cat. No.
    MANA528011
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Filtration speed
    22 s
  • MN 715
    125 mm
    1 * 100 Items
    1 * 100 Items - 311,00 kr
    311,00 kr
    1 * 100 Items
       311,00 kr
    Specifications
    Cat. No.
    MANA528012
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Filtration speed
    22 s
  • MN 715
    150 mm
    1 * 100 Items
    1 * 100 Items - 311,00 kr
    311,00 kr
    1 * 100 Items
       311,00 kr
    Specifications
    Cat. No.
    MANA528015
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Filtration speed
    22 s
  • MN 715
    185 mm
    1 * 100 Items
    1 * 100 Items - 344,00 kr
    344,00 kr
    1 * 100 Items
       344,00 kr
    Specifications
    Cat. No.
    MANA528018
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Filtration speed
    22 s
  • MN 715
    240 mm
    1 * 100 Items
    1 * 100 Items - 508,00 kr
    508,00 kr
    1 * 100 Items
       508,00 kr
    Specifications
    Cat. No.
    MANA528024
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Filtration speed
    22 s
  • MN 715
    270 mm
    1 * 100 Items
    1 * 100 Items - 587,00 kr
    587,00 kr
    1 * 100 Items
       587,00 kr
    Specifications
    Cat. No.
    MANA528027
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Filtration speed
    22 s
  • MN 715
    320 mm
    1 * 100 Items
    1 * 100 Items - 1.330,00 kr
    1.330,00 kr
    1 * 100 Items
       1.330,00 kr
    Specifications
    Cat. No.
    MANA561032
    Weight
    70 g/m²
    Thickness
    0,16 mm
    Filtration speed
    22 s