Fmoc-Osu (N-(9-fluorenylmethoxycarbonyloxy)succinimide) ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
Synonyms: Fmoc-OSu, N-(9-Fluorenylmethoxycarbonyloxy)succinimide
CAS Number:82911-69-1
:
Formula:C₁₉H₁₅NO₅
MDL Number:MFCD00010733
:337.3 g/mol
Melting Pt:153°C
Storage Temperature:Refrigerator
EINECS:433-520-5
UN:3077
ADR:9,III
:GHS07
:GHS08
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Butylamine Method):min. 98.0 %
Specification Test Results
Related Information
Frequently Bought Together