
Sulphosuccinic acid ≥70% in aqueous solution for synthesis, Sigma-Aldrich®
Katalog # 8.14413.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 ml
- Seq:2440
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Boiling Point:140 °C (1013 hPa)
- Density:1.46 g/cm3 (20 °C)
- pH:1 (H2O, 20 °C) (saturated aqueous solution)
Specification Test Results
About this item
:
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:225-898-3
UN:3265
:GHS05
Storage Temperature:Ambient
CAS Number:5138-18-1
Formula:C₄H₆O₇S
MDL Number:MFCD00075139