
Creatine (N-amidinosarcosine) hydrate ≥98.0% (by HPLC, titration analysis)
: TCI
:6020-87-7
:
:C₄H₉N₃O₂·xH₂O
:MFCD00004282
:131.14 g/mol (anhy)
:14,02568
:200-306-6
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0% (calcd.on dried substance)
- Solubility in hot Water:almost transparency
- Drying loss:6.0 to 13.0 %
Frequently Bought Together