Order Entry
Denmark
ContactUsLinkComponent
 
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG) ≥98.0% (by HPLC) substrate for beta-D-Galactosidase
 :  TCI
 
undefined
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG) ≥98.0% (by HPLC) substrate for beta-D-Galactosidase
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystaline
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 97.0 %
  • Specific rotation [a]20/D:
    -66.0 to -71.5 deg (C=1, H2O)
  • beta-Galactosidase detection test:
    to pass test

 

Frequently Bought Together