Order Entry
Denmark
ContactUsLinkComponent
Titanium(IV) chloride 14% ∼1.0 mol/L in dichloromethane
Corrosive
Toxic
Health
Environment
Titanium(IV) chloride 14% ∼1.0 mol/L in dichloromethane
Supplier:  TCI
Certificates
undefined
Corrosive
Toxic
Health
Environment
Titanium(IV) chloride 14% ∼1.0 mol/L in dichloromethane
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Red to Green clear liquid
  • Concentration (Argentmetric titration):
    14.0 to 15.0 w/w%

Specification Test Results

Related Information

Frequently Bought Together