Broxyquinoline ≥98.0% (by titrimetric analysis)
TCIAD0208-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
- Appearance:White to Light yellow to Green powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:197.0 to 203.0 deg-C
- Solubility in THF:almost transparency
: 5,7-Dibromo-8-hydroxyquinoline, 5,7-Dibromo-8-quinolinol
:
:302.95 g/mol
:208-317-8
:GHS07
:Ambient
:521-74-4
:C₉H₅Br₂NO
:MFCD00006785
:371
:14,01453