Order Entry
Denmark
ContactUsLinkComponent
Toluene ≥99.5% (by GC) for spectrophotometry
Flammable
Irritant
Health
Toluene ≥99.5% (by GC) for spectrophotometry
Catalog # TCIAT0260-100ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
Health
Toluene ≥99.5% (by GC) for spectrophotometry
Catalog # TCIAT0260-100ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.5 %
  • Absorbance (290nm):
    max. 0.350
  • Absorbance (300nm):
    max. 0.150
  • Absorbance (310nm):
    max. 0.080
  • Absorbance(300nm):
    max. 0.150
  • Absorbance(310nm):
    max. 0.080
  • Absorbance(320nm):
    max. 0.050
  • Absorbance(330nm):
    max. 0.020
  • Absorbance(340nm):
    max. 0.010
  • Absorbance(400nm):
    max. 0.010
  • Water:
    max. 0.03 %

Specification Test Results

About this item

 :
 :92.14 g/mol
Merck Index:14,09529
Boiling Pt:110°C (lit.)
Melting Pt:-95°C (lit.)
Melting Pt:–95°C (lit.)
Density:0,867 g/cm³ (20 °C)
Flash Pt:5°C (lit.)
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
REACH:05-2114615130-69
 :GHS02
 :GHS07
 :GHS08
CAS Number:108-88-3
Formula:C₆H₅CH₃
MDL Number:MFCD00008512