n-Tridecane ≥99.0% (by GC)
: TCI
: Tridecane
:629-50-5
:
:H₃C(CH₂)₁₁CH₃
:MFCD00008979
:235°C
:184.37 g/mol
:–5°C
:79°C
:211-093-4
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %
Frequently Bought Together