Order Entry
Denmark
ContactUsLinkComponent
 
 
 
Diethyl ether, anhydrous ≥99.5% (by GC) stabilised
  TCIAD3479-500ML
 :  TCI
 :  
undefined
 
 
Diethyl ether, anhydrous ≥99.5% (by GC) stabilised
  TCIAD3479-500ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

 

 

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.5 %
  • Water:
    max. 50 ppm

 

 

 :
 :74.12 g/mol
 :14,03806
 :35°C
 :–116°C
 :0,7135 g/cm³ (20 °C)
 :–45°C
 :Ambient
 :200-467-2
 :1155
 :3,I
 :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
 :01-2119535785-29
 :GHS02
 :GHS07
 :60-29-7
 :(CH₃CH₂)₂O
 :MFCD00011646