Order Entry
Denmark
ContactUsLinkComponent
N,O-Bis(trimethylsilyl)trifluoroacetamide ≥95.0% (by GC) for gas chromatography
Flammable
Corrosive
N,O-Bis(trimethylsilyl)trifluoroacetamide ≥95.0% (by GC) for gas chromatography
Catalog # TCIAA5603-5ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Corrosive
N,O-Bis(trimethylsilyl)trifluoroacetamide ≥95.0% (by GC) for gas chromatography
Catalog # TCIAA5603-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 95.0 %
  • High boiling impurity of Silylation agent:
    No impurity higher than Isopropyl myristate (50 ppm) after 5 min + hold up time
  • Effect test of TMS derivatization:
    Effective as derivatization agent DL-Valine

Specification Test Results

About this item

 :
 :257.4 g/mol
Boiling Pt:142°C
Melting Pt:–41 °C
Density:0,96 g/cm³ (20 °C)
Flash Pt:24°C
Storage Temperature:Refrigerator
EINECS:247-103-9
UN:2924
ADR:3,III
 :GHS02
 :GHS05
CAS Number:25561-30-2
Formula:C₈H₁₈F₃NOSi₂
MDL Number:MFCD00008269