Manganese(III) acetylacetonate ≥98.0% (by titrimetric analysis)
: TCI
: Manganese (III) tris(2,4-pentanedionato-O,O'), Tris(2,4-pentanedionato-O,O')manganese (III), Manganese (III) 2,4-pentanedionate
:14284-89-0
:
:C₁₅H₂₁MnO₆
:MFCD00000023
:352.27 g/mol
:160
:238-188-3
:GHS07
- Environmentally Preferable:
- Appearance:Black powder to crystal
- Purity (Chelometric Titration):min. 98.0 %
- Solubility in dilute HCl:almost transparency
Frequently Bought Together