Order Entry
Denmark
ContactUsLinkComponent
3,5-Dihydroxytoluene, anhydrous ≥98.0% (by GC)
Irritant
3,5-Dihydroxytoluene, anhydrous ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
3,5-Dihydroxytoluene, anhydrous ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystaline
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    108.0 to 111.0 deg-C
  • Solubility in Methanol:
    very faint turbidity

Specification Test Results

Related Information

Frequently Bought Together