Order Entry
Denmark
ContactUsLinkComponent
1,3-Cyclohexanediol cis- and trans-mixture ≥98.0% (by GC)
1,3-Cyclohexanediol cis- and trans-mixture ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
1,3-Cyclohexanediol cis- and trans-mixture ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0% (total of isomer)

Specification Test Results

Related Information

Frequently Bought Together