Order Entry
Denmark
ContactUsLinkComponent
 
 
Tetrabutylammonium chloride ≥98.0% (by titrimetric analysis) for ion pair chromatography
 :  TCI
 
undefined
 
Tetrabutylammonium chloride ≥98.0% (by titrimetric analysis) for ion pair chromatography
 :  TCI
 

 

  • Appearance:
    White powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Absorbance (230nm):
    max. 0.080
  • Absorbance (254nm):
    max. 0.040
  • Absorbance (400nm):
    max. 0.010

 

Frequently Bought Together