2-Ethylpyrazine
Glass bottle for solids, 1 g
Catalog # ROTH5720.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Ethylpyrazine
- CAS Number:13925-00-3
- Formula:C₆H₈N₂
- MDL Number:MFCD00006149
- Molecular Weight:108.14 g/mol
- Boiling Pt:152 °C (1013 hPa)
- Density:0.98 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:237-691-5
- UN:1993
- ADR:3,III
- :Density: 0.98 g/cm³ (20 °C)
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



