Solid phase extraction cartridges, Bond Elut™ C18 OH
: VARIAN
Bond Elut™ C18 OH is a non-end capped version of the octadecyl bonded phases that enables the silanols on the silica surface to be more active. This low-load C18 has well-controlled silanol activity that permits the fractionation of metabolites and enhances retention of basic compounds compared to an endcapped C18. The pore size makes it a good choice for peptide extraction.
- Silanol activity permits metabolite fractionation
- Tight QC tolerances deliver batch-to-batch reproducibility
- 150 Å pore size expands retention window to higher molecular weight compounds
- Catalog No:
- VARI12102065
- VARI12102020
- VARI12102046
- VARI12256040
- Carbon load:
- 14,9%
- 14,9%
- 14,9%
- 14,9%
- Endcapped:
- No
- No
- No
- No
- Surface area:
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
Frequently Bought Together