Order Entry
Denmark
ContactUsLinkComponent
Eriochrome cyanine R, Supelco®
Eriochrome cyanine R, Supelco®
Catalog # 1.03164.0025
Supplier:  Merck
CAS Number:  
Eriochrome cyanine R, Supelco®
Catalog # 1.03164.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Solubility:
    420 g/l (20 °C)
  • Molar Mass:
    536.4 g/mol
  • Bulk Density:
    590 kg/m3
  • pH:
    9.1 (10 g/l, H2O, 24 °C)

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ200
EINECS:222-641-7
CAS Number:3564-18-9
Formula:C₂₃H₁₅Na₃O₉S
MDL Number:MFCD00012411