Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, 5α-Cholestane 10000 µg/ml in chloroform, Supelco®
Organic reference standard, 5α-Cholestane 10000 µg/ml in chloroform, Supelco®
  47124.
 :  Merck
 :  
undefined
Organic reference standard, 5α-Cholestane 10000 µg/ml in chloroform, Supelco®
  47124.
 :  Merck
 :  

 

  • Pk:
    1
  • CAS No.:
    481-21-0
  • Description:
    5α-Cholestane 10,000 µg/ml in chloroform
  • Environmentally Preferable:
  • Formula:
    C₂₇H₄₈
  • Description:
    5α-Cholestane