Organic reference standard, (R)-(+)-Cathinone hydrochloride 1000 µg/ml in methanol, Cerilliant
CERIC-028
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:71031-15-7
- Description:(R)-(+)-Cathinone hydrochloride 1,000 µg/ml in methanol
- Environmentally Preferable:
- Formula:C₉H₁₁NO
- Description:(R)-(+)-Cathinone hydrochloride