Order Entry
Denmark
ContactUsLinkComponent
 
Magnesium 8-anilinonaphthalene-1-sulphonate hydrate ≥98.0% (by HPLC, titration analysis) hydrophobic fluorescent probe
Catalog # TCIAA5353-1G
Supplier:  TCI
CAS Number:  
undefined
Magnesium 8-anilinonaphthalene-1-sulphonate hydrate ≥98.0% (by HPLC, titration analysis) hydrophobic fluorescent probe
Catalog # TCIAA5353-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Chelometric Titration):
    min. 98.0 % (calcd.on anh.substance)
  • Water:
    max. 10.0 %

Specification Test Results

About this item

 :
 :620.98 g/mol (anhy)
EINECS:242-004-7
 :GHS07
CAS Number:18108-68-4
Formula:C₃₂H₂₄MgN₂O₆S₂·xH₂O
MDL Number:MFCD00065353