
Chromotrope 2R
Catalog # TCIAC0547-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Red to Dark blue to Black powder to crystal
- Lambda max.:508.0 to 512.0 nm (MeOH)
- Absorbance (E1%1cm):min. 350 (MeOH, 508.0 to 512.0 nm)
Specification Test Results
About this item
:
:468.37 g/mol
EINECS:224-085-0
:GHS07
CAS Number:4197-07-3
Formula:C₁₆H₁₀N₂Na₂O₈S₂
MDL Number:MFCD00003955