Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

β-Cyclodextrine ≥98.0% (by HPLC)

Supplier:   TCI
β-Cyclodextrine ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
β-Cyclodextrine ≥98.0% (by HPLC)

3 Options Available Below

Product Details & Documents
Synonyms:  Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
  • CAS Number:7585-39-9
  • Formula:C₄₂H₇₀O₃₅
  • MDL Number:MFCD00078139
  • Molecular Weight:1134.98 g/mol
  • Merck Index:14,02718
  • EINECS:231-493-2
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Specific rotation [a]20/D:
    +160 to +166 deg (C=1, H2O) (calcd.on anh.substance)
  • Water:
    max. 14.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 165,00 kr
    165,00 kr
    1 * 25 g
       165,00 kr

  • 100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 280,00 kr
    280,00 kr
    1 * 100 g
       280,00 kr

  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 714,00 kr
    714,00 kr
    1 * 500 g
       714,00 kr