
2-Imidazolidinone ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:120-93-4
:
Formula:C₃H₆N₂O
MDL Number:MFCD00005257
:86.0925 g/mol
Merck Index:14,04914
EINECS:204-436-4
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:131.0 to 136.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together