

Bis(2-chloroethyl)ammonium chloride ≥99.0% (by titrimetric analysis)
: TCI
:821-48-7
:
:C₄H₁₀Cl₃N
:MFCD00012515
:178.49 g/mol
:212-479-5
:8,II
:GHS07
:GHS08
- Appearance:White to Light yellow to Light orange powder to crystaline
- Purity (Argentmetric Titration):min. 99.0 %
- Solubility in Water:almost transparency
Frequently Bought Together