Order Entry
Denmark
ContactUsLinkComponent
4,4''-Diiodo-p-terphenyl ≥98.0% (by GC)
Corrosive
Environment
4,4''-Diiodo-p-terphenyl ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
Corrosive
Environment
4,4''-Diiodo-p-terphenyl ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %

Specification Test Results

Frequently Bought Together